Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR3/TNFRSF10C Antibody (003), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289484C
Description
TRAILR3/TNFRSF10C Monoclonal specifically detects TRAILR3/TNFRSF10C in Human samples. It is validated for Flow Cytometry.Specifications
TRAILR3/TNFRSF10C | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
3 | |
Flow Cytometry | |
Antagonist decoy receptor for TRAIL/Apo-2L, CD263, CD263 antigen, cytotoxic TRAIL receptor-3, DcR1, DCR1MGC149502, Decoy receptor 1, Decoy TRAIL receptor without death domain, LITMGC149501, Lymphocyte inhibitor of TRAIL, TNFRSF10C, TRAIL receptor 3, TRAIL receptor without an intracellular domain, TRAIL-R3, TRAILR3TNF-related apoptosis-inducing ligand receptor 3, TRIDDCR1-TNFR, tumor necrosis factor receptor superfamily member 10C, tumor necrosis factor receptor superfamily, member 10c, decoy without anintracellular domain | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAILR3/TNFRSF10C (NP_003832.2; Met1-Pro235). | |
0.1 mL | |
Apoptosis, Breast Cancer, Cell Biology, Chromatin Research, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
8794 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction