Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR3/TNFRSF10C Antibody (90906), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6302Z
This item is not returnable.
View return policy
Description
TRAILR3/TNFRSF10C Monoclonal antibody specifically detects TRAILR3/TNFRSF10C in Human samples. It is validated for Flow CytometrySpecifications
TRAILR3/TNFRSF10C | |
Monoclonal | |
DyLight 755 | |
Antagonist decoy receptor for TRAIL/Apo-2L, CD263, CD263 antigen, cytotoxic TRAIL receptor-3, DcR1, DCR1MGC149502, Decoy receptor 1, Decoy TRAIL receptor without death domain, LITMGC149501, Lymphocyte inhibitor of TRAIL, TNFRSF10C, TRAIL receptor 3, TRAIL receptor without an intracellular domain, TRAIL-R3, TRAILR3TNF-related apoptosis-inducing ligand receptor 3, TRIDDCR1-TNFR, tumor necrosis factor receptor superfamily member 10C, tumor necrosis factor receptor superfamily, member 10c, decoy without anintracellular domain | |
Mouse myeloma cell line NS0-derived recombinant human TRAIL R3/TNFRSF10C, Met1-Ala221, Accession # O14798 | |
0.5 mL | |
Apoptosis, Breast Cancer, Cell Biology, Chromatin Research, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
8794 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry | |
90906 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction