Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR4/TNFRSF10D/DcR2 Antibody (104918), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB633L
This item is not returnable.
View return policy
Description
TRAILR4/TNFRSF10D/DcR2 Monoclonal antibody specifically detects TRAILR4/TNFRSF10D/DcR2 in Human samples. It is validated for Flow Cytometry, ELISA (Matched Antibody Pair)Specifications
TRAILR4/TNFRSF10D/DcR2 | |
Monoclonal | |
DyLight 550 | |
CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain | |
Mouse myeloma cell line NS0-derived recombinant human TRAIL R4/TNFRSF10D, Ala56-His211, Accession # Q9UBN6 | |
0.5 mL | |
Apoptosis, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
8793 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, ELISA | |
104918 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction