Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR4/TNFRSF10D/DcR2 Antibody (201), DyLight 488

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289481G
Description
TRAILR4/TNFRSF10D/DcR2 Monoclonal specifically detects TRAILR4/TNFRSF10D/DcR2 in Human samples. It is validated for Western Blot.Specifications
TRAILR4/TNFRSF10D/DcR2 | |
Monoclonal | |
DyLight 488 | |
CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAILR4/TNFRSF10D/DcR2 (NP_003831.2; Met1-His211). | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
8793 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
201 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction