Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAILR4/TNFRSF10D/DcR2 Antibody (MM0579-6G9), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211931AF700
Description
TRAILR4/TNFRSF10D/DcR2 Monoclonal specifically detects TRAILR4/TNFRSF10D/DcR2 in Human samples. It is validated for Western Blot.Specifications
TRAILR4/TNFRSF10D/DcR2 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TNFRSF10D | |
Human recombinant TRAILR4/TNFRSF10D/DcR2 (extracellular domain) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
MM0579-6G9 | |
Western Blot, Flow Cytometry | |
CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain | |
Mouse | |
Protein G purified | |
Apoptosis, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity | |
8793 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction