Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAIP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
| Antigen | TRAIP |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TRAIP Polyclonal specifically detects TRAIP in Human, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TRAIP | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Xenopus species | |
| RNF206TRIPRING finger protein 206, TRAF interacting protein, TRAF-interacting protein | |
| TRAIP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 10293 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QSQRPEVEEMIRDMGVGQSAVEQLAVYCVSLKKEYENLKEARKASGEVADKLRKDLFSSRSKLQTVYSELDQAKLELKSAQKDLQSADKEIMSLK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title