Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAIP Antibody (OTI2D4), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274580V
Description
TRAIP Monoclonal specifically detects TRAIP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
TRAIP | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TRAIP | |
Recombinant protein fragment corresponding to amino acids 67-313 of human TRAIP (NP_005870) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI2D4 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
RNF206TRIPRING finger protein 206, TRAF interacting protein, TRAF-interacting protein | |
Mouse | |
Affinity Purified | |
Zinc Finger | |
10293.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction