Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAIP Monoclonal Antibody (OTI3A4), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

$516.29

Specifications

Antigen TRAIP
Clone OTI3A4
Concentration 1 mg/mL
Applications Flow Cytometry, Western Blot
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-167-8072
SDP
Encompass_Preferred
View Documents
OriGene
TA800087
100 μL
Each of 1 for $516.29
Only null left
Add to Cart
 
Description

Description

TRAIP Monoclonal antibody specifically detects TRAIP in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

TRAIP
1 mg/mL
Monoclonal
Liquid
RUO
Q9BWF2
IgG1
-20° C, Avoid Freeze/Thaw Cycles
OTI3A4
Flow Cytometry, Western Blot
Unconjugated
Mouse
Human
10293
Primary
TRAIP
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.