Learn More
TRAIP Mouse anti-Human, Clone: OTI1G3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800090

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.Specifications
TRAIP | |
Monoclonal | |
Unconjugated | |
TRAIP | |
RNF206, SCKL9, TRIP | |
Mouse | |
Affinity Chromatography | |
RUO | |
10293 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI1G3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9BWF2 | |
TRAIP | |
Human recombit protein fragment corresponding to amino acids 67-313 of human TRAIP produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.