Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564938 Supplier Invitrogen™ Supplier No. PA564938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CTPRRLSLAES FTNTRESTTT MSTSLGLVWL LKERGISAAV YDPQSWDRAG RGSLLHSYTP KMAVIPSTPP NSPMQTPTSS PPSFEFKCTS PPYDNFLASK PASSILREVR EKNVRSSESQ TDVSVSNLNL VD Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

Involved in the regulation of endosome-to-lysosome trafficking, including endocytic trafficking of EGF-EGFR complexes and GABA-A receptors. Involved in mitochondrial motility. When O-glycosylated, abolishes mitochondrial motility. Crucial for recruiting OGT to the mitochondrial surface of neuronal processes. TRAK1 and RHOT form an essential protein complex that links KIF5 to mitochondria for light chain-independent, anterograde transport of mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAK1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRAK1
Gene Accession No. Q9UPV9
Gene Alias 106 kDa O-GlcNAc transferase-interacting protein; 2310001H13Rik; AI413908; AI467545; hyrt; KIAA1042; MILT1; milton homolog 1; mKIAA1042; OGT(O-Glc-NAc transferase)-interacting protein 106 KDa; OIP106; O-linked N-acetylglucosamine transferase interacting protein 106; Protein Milton; RGD1307844; trafficking kinesin protein 1; trafficking kinesin-binding protein 1; trafficking protein, kinesin binding 1; TRAK1
Gene Symbols TRAK1
Host Species Rabbit
Immunogen Recombinant Human TRAK1. Recombinant protein control fragment (Product #RP-105720).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.