Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAK2 Monoclonal Antibody (S390-43), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA527606

Catalog No. PIMA527606


Description

Description

May regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRAK2
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
O60296, Q8R2H7
Trak2
Fusion protein amino acids 757-914 (C-terminus) of human TRAK2
100 μg
Primary
Human, Rat
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S390-43
Unconjugated
TRAK2
Trafficking kinesin-binding protein 2, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, GABA-A receptor-interacting factor 1, O-GlcNAc transferase-interacting protein of 98 kDa, EMBL AAH60681.1, Ensembl ENSMUSP00000027186 MGI 1918077, Als2cr3, Grif1, Oip98, KIAA0549, TRAK2_Human
Mouse
Protein G
RUO
171086, 66008
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.