Learn More
Invitrogen™ TRAK2 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TRAK2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TRAK2 |
| Gene Accession No. | O60296 |
| Gene Alias | 2900022D04Rik; 4733401O11Rik; AI480836; Als2cr3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3 homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein homolog; AW046271; CALS-C; EMBL AAH60681.1; GABA-A receptor interacting factor-1; GABA-A receptor-interacting factor 1; gamma-aminobutyric acid(A) receptor-interacting factor; Grif1; GRIF-1; KIAA0549; MILT2; milton homolog 2; mKIAA0549; O-GlcNAc transferase-interacting protein of 98 kDa; OIP98; O-linked N-acetylglucosamine transferase interacting protein 98; trafficking kinesin protein 2; trafficking kinesin-binding protein 2; trafficking protein, kinesin binding 2; Trak2 |
| Gene Symbols | TRAK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.