Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104181
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104181 Supplier Invitrogen™ Supplier No. PA5104181
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TRAK2.

May regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRAK2
Gene Accession No. O60296
Gene Alias 2900022D04Rik; 4733401O11Rik; AI480836; Als2cr3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3 homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein homolog; AW046271; CALS-C; EMBL AAH60681.1; GABA-A receptor interacting factor-1; GABA-A receptor-interacting factor 1; gamma-aminobutyric acid(A) receptor-interacting factor; Grif1; GRIF-1; KIAA0549; MILT2; milton homolog 2; mKIAA0549; O-GlcNAc transferase-interacting protein of 98 kDa; OIP98; O-linked N-acetylglucosamine transferase interacting protein 98; trafficking kinesin protein 2; trafficking kinesin-binding protein 2; trafficking protein, kinesin binding 2; Trak2
Gene Symbols TRAK2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRAK2(Accession O60296), corresponding to amino acid residues A861-L911.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66008
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.