Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRAK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | TRAK2 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111544
|
Invitrogen™
PA5111544 |
100 μL |
Each of 1 for $589.50
|
|
|||||
Description
Immunogen sequence: DEEEGITFQV QQPLEVEEKL STSKPVTGIF LPPITSAGGP VTVATANPGK CLSCTNST.
May regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.Specifications
TRAK2 | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60296 | |
66008 | |
Recombinant Protein corresonding to Human TRAK2. Recombinant protein control fragment (Product #RP-107138). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
TRAK2 | |
2900022D04Rik; 4733401O11Rik; AI480836; Als2cr3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3 homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein homolog; AW046271; CALS-C; EMBL AAH60681.1; GABA-A receptor interacting factor-1; GABA-A receptor-interacting factor 1; gamma-aminobutyric acid(A) receptor-interacting factor; Grif1; GRIF-1; KIAA0549; MILT2; milton homolog 2; mKIAA0549; O-GlcNAc transferase-interacting protein of 98 kDa; OIP98; O-linked N-acetylglucosamine transferase interacting protein 98; trafficking kinesin protein 2; trafficking kinesin-binding protein 2; trafficking protein, kinesin binding 2; Trak2 | |
TRAK2 | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title