Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554931 Supplier Invitrogen™ Supplier No. PA554931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRWREHSAFQ APAVKKKPTV TKGRSSKKGT ENGVNGTLTS NVADSPRNK Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.

TRAM, or TRIF-related adaptor molecule, plays an essential role in the MyD88-independent signaling of TLR4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tram1
Gene Accession No. Q15629
Gene Alias 1810049E02Rik; PNAS8; TRAM; Tram1; TRAMP; Translocating chain-associated membrane protein 1; translocating chain-associating membrane protein; translocating chain-associating membrane protein 1; translocation associated membrane protein 1; translocation-associating membrane protein 1
Gene Symbols Tram1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRAM1. Recombinant protein control fragment (Product #RP-93737).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23471
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.