Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580181
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580181 Supplier Invitrogen™ Supplier No. PA580181
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Kidney Tissue, 293T Cell, RAJI Cell, JURKAT Cell, Hela Cell. IHC: Human Lung Cancer Tissue, Rat Kidney Tissue IHC-F: rat intestine tissue, rat brain tissue. ICC/IF: HELA Cell.

TRAM, or TRIF-related adaptor molecule, plays an essential role in the MyD88-independent signaling of TLR4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAM1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Tram1
Gene Accession No. Q15629, Q5XI41, Q91V04
Gene Alias 1810049E02Rik; PNAS8; TRAM; Tram1; TRAMP; Translocating chain-associated membrane protein 1; translocating chain-associating membrane protein; translocating chain-associating membrane protein 1; translocation associated membrane protein 1; translocation-associating membrane protein 1
Gene Symbols Tram1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human TRAM1 (314-331aa KFINFQLRRWREHSAFQA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23471, 312903, 72265
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.