Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590249
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590249 Supplier Invitrogen™ Supplier No. PA590249
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CVTQAFMMWK FINFQLRRWR EHSAFQAPAV KKKPTVTKGR SSKKGTENGV NGTLTSNVAD SPRNKKEKSS; Positive Samples: U-251MG, mouse liver; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.

TRAM, or TRIF-related adaptor molecule, plays an essential role in the MyD88-independent signaling of TLR4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Tram1
Gene Accession No. Q15629, Q91V04
Gene Alias 1810049E02Rik; PNAS8; TRAM; Tram1; TRAMP; Translocating chain-associated membrane protein 1; translocating chain-associating membrane protein; translocating chain-associating membrane protein 1; translocation associated membrane protein 1; translocation-associating membrane protein 1
Gene Symbols Tram1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 305-374 of human TRAM1 (NP_0551091).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23471, 72265
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.