Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAM1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3996 Supplier Bioss Supplier No. BS16565R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRAM, or TRIF-related adaptor molecule, plays an essential role in the MyD88-independent signaling of TLR4.

Specifications

Antigen TRAM1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Tram1
Gene Accession No. Q15629
Gene Alias 1810049E02Rik; PNAS8; TRAM; Tram1; TRAMP; Translocating chain-associated membrane protein 1; translocating chain-associating membrane protein; translocating chain-associating membrane protein 1; translocation associated membrane protein 1; translocation-associating membrane protein 1
Gene Symbols Tram1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TRAM1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23471
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.