Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRAM1L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560750
Description
Immunogen sequence: WLQRWVEDSN IQASCMKKKR SRSSK Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 60%.
TRAM1L1 is stimulatory or required for the translocation of secretory proteins across the ER membrane.
Specifications
TRAM1L1 | |
Polyclonal | |
Unconjugated | |
TRAM1L1 | |
TRAM1L1; Translocating chain-associated membrane protein 1-like 1; translocation associated membrane protein 1 like 1; translocation associated membrane protein 1-like 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
133022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N609 | |
TRAM1L1 | |
Recombinant protein corresponding to Human TRAM1L1. Recombinant protein control fragment (Product #RP-96743). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction