Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAM2 Antibody (SR1741), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32163850UL
Description
TRAM2 Monoclonal antibody specifically detects TRAM2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TRAM2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
9697 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot | |
SR1741 | |
Western Blot 1:500-1:2000 | |
KIAA0057TRAM-like protein, translocating chain-associated membrane protein 2, translocation associated membrane protein 2 | |
A synthesized peptide derived from human TRAM2 (Uniprot #: Q15035) | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction