Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563503
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563503 Supplier Invitrogen™ Supplier No. PA563503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFLFILPQYN ISVPTADSET VHYHYGPKDL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 90%.

Necessary for collagen type I synthesis. May couple the activity of the ER Ca(2+) pump SERCA2B with the activity of the translocon. This coupling may increase the local Ca(2+) concentration at the site of collagen synthesis, and a high Ca(2+) concentration may be necessary for the function of molecular chaperones involved in collagen folding. Required for proper insertion of the first transmembrane helix N-terminus of TM4SF20 into the ER lumen, may act as a ceramide sensor for regulated alternative translocation (RAT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAM2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRAM2
Gene Accession No. Q15035
Gene Alias C330003D03Rik; KIAA0057; LOC100361830; tram2; TRAM-like protein; Translocating chain-associated membrane protein 2; translocating chain-associating membrane protein 2; translocation associated membrane protein 2; wu:fb20c11; wu:fi23a12; zgc:111902; zgc:55869; zgc:76937
Gene Symbols TRAM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRAM2. Recombinant protein control fragment (Product #RP-103312).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9697
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.