Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Trametinib
GREENER_CHOICE

Catalog No. 770910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-091-0 10 mg
77-095-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 77-091-0 Supplier Tocris Bioscience™ Supplier No. 7709/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective MEK inhibitor; orally bioavailable

Trametinib is a potent and selective MEK1 and MEK2 inhibitor (IC50 = 0.92 and 1.8 nM, respectively, in cell-free assays). In vitro, it inhibits the growth of human colorectal cancer cell lines expressing either B-RAF or K-RAF (IC50 values range from 0.48 to 36 nM). In vivo, it suppresses tumor growth of HT-29 and COLO205 xenografts in nude mice. Trametinib induces apoptosis. This compound is orally bioavailable.

Specifications

Chemical Name or Material N-[3-[3-Cyclopropyl-5-[(2-fluoro-4-iodophenyl)amino]-3,4,6,7-tetrahydro-6,8-dimethyl-2,4,7-trioxopyrido[4,3-d]pyrimidin-1(2 H)-yl]phenyl]acetamide
CAS 871700-17-3
Quantity 10 mg
Target MEK Inhibitors
Molecular Formula C26H23FIN5O4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.