Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody (101), Alexa Fluor™ 700

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289957AF700
Description
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA Detection.Specifications
TRANCE/TNFSF11/RANK L | |
Monoclonal | |
Alexa Fluor 700 | |
CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human TRANCE/TNFSF11/RANK L (O14788-2; Gly63-Asp244). | |
0.1 mL | |
Cancer | |
8600 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
101 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction