Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRANCE/TNFSF11/RANK L Rabbit anti-Human, Clone: R001, Novus Biologicals™

Rabbit Monoclonal Antibody
$524.00 - $707.00
Specifications
Antigen | TRANCE/TNFSF11/RANK L |
---|---|
Clone | R001 |
Applications | Neutralization |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human samples. It is validated for Neutralization.Specifications
TRANCE/TNFSF11/RANK L | |
Neutralization | |
Unconjugated | |
Rabbit | |
Cancer | |
0.2 um filtered solution in Histidine and Arginine buffer, 120mM NaCl, 0.02% Tween 80, pH6.0 | |
8600 | |
IgG | |
Protein A purified |
R001 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
Recombinant Human TRANCE/TNFSF11/RANK L Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title