Alfa Aesar™ trans-2,3-Epoxybutane, 97%

Manufacturer: Alfa Aesar™ B2200514

Quantity 25g

 View more versions of this product

Catalog No. AAB2200514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21490-63-1
    Molecular Formula C4H8O
    Formula Weight 72.11
    MDL Number MFCD00005129
    UN Number UN3271
    Beilstein 79772
    Synonym trans-beta-Butylene oxide
    Chemical Name or Material trans-2,3-Epoxybutane

    Specifications

    Formula Weight 72.11
    Boiling Point 54° to 55°C
    Odor Recognizable
    Flash Point −26°C (−14°F)
    Molecular Formula C4H8O
    MDL Number MFCD00005129
    CAS 21490-63-1
    UN Number UN3271
    Beilstein 79772
    Synonym trans-beta-Butylene oxide
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H341-H315-H319-H335
    Highly flammable liquid and vapour.
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 25g
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
    RTECS Number EK3855040
    Chemical Name or Material trans-2,3-Epoxybutane
    Assay 97%

    Description

    Applications
    A higher order cuprate reacted with trans-2,3-epoxybutane a high yield both counted on the epoxide (96%) and on the haloalkane (85%). Enolate derived from trans-2,3-Epoxybutan has chiral recognition.

    Solubility
    Soluble in water(95g/L).

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

    GHS H Statement
    H225-H341-H315-H319-H335
    Highly flammable liquid and vapour.
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit