Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline 95.0+%, TCI America™
SDP

Catalog No. D46521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46521G 1 g
D4652200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46521G Supplier TCI America Supplier No. D46521G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 908294-68-8
Molecular Formula C18H22N2
Molecular Weight (g/mol) 266.39
MDL Number MFCD06669887
InChI Key BABQFMPURPKJNW-UHFFFAOYSA-N
PubChem CID 1284716
IUPAC Name N-[2,6-bis(propan-2-yl)phenyl]-1-(pyridin-2-yl)methanimine
SMILES CC(C)C1=CC=CC(C(C)C)=C1N=CC1=CC=CC=N1

Specifications

Quantity 1 g
Formula Weight 266.39
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,6-Diisopropyl-N-(2-pyridylmethylene)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.