Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hepten-1-ol 96.0+%, TCI America™
SDP

Catalog No. H068225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H068225ML 25 mL
H06825ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H068225ML Supplier TCI America Supplier No. H068225ML

Chemical Identifiers

CAS 33467-76-4
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00014056
InChI Key ASFYPVGAALGVNR-AATRIKPKSA-N
Synonym trans-2-hepten-1-ol, e-2-hepten-1-ol, e-hept-2-en-1-ol, 2-hepten-1-ol, e, 2-hepten-1-ol, 2e, 2-heptenol, unii-q989yfo10i, 2-hepten-1-ol, hept-2-en-1-ol, 2e-hept-2-en-1-ol
PubChem CID 5318017
IUPAC Name (E)-hept-2-en-1-ol
SMILES CCCCC=CCO

Specifications

Color Colorless
Boiling Point 75°C
Quantity 25 mL
UN Number 1987
Formula Weight 114.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hepten-1-ol
EINECSNumber (2)-2393
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.