Learn More
trans-2-Hexenal, 96%, Thermo Scientific Chemicals
Description
trans-2-Hexenal is used in evaluation of quality of protected designation of origin virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. It was used in the multicomponent method for the determination of low-molecular-weight carbonyl compounds in biological samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-2-Hexenal is used in evaluation of quality of protected designation of origin virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis. It was used in the multicomponent method for the determination of low-molecular-weight carbonyl compounds in biological samples.
Solubility
Not miscible in water.
Notes
Store at 4°C. Incompatible materials are bases, air, oxidizing agents, reducing agents and heat.
Specifications
Density | 0.848 |
Boiling Point | 37°C to 38°C (8 mmHg) |
Flash Point | 38°C (100°F) |
Odor | Sweet |
Refractive Index | 1.447 |
Quantity | 25 g |
UN Number | UN1988 |
Beilstein | 1699684 |
Sensitivity | Air sensitive |
Solubility Information | Not miscible in water. |
Show More |
Safety and Handling
Hazard Category | H226-H302-H311-H317-H335 |
Hazard Statement | GHS H Statement H311-H226-H302-H332-H315-H319-H335 Toxic in contact with skin. Flammable liquid and vapor. Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P333+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, FLAMMABLE, TOXIC, N.O.S. |
EINECSNumber | 229-778-1 |
RTECSNumber | MP5900000 |
TSCA | Yes |
Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.