Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Hexenyl acetate, 98%

Catalog No. AAA1878630
Click to view available options
Quantity:
50 g
250 g

CAS: 2497-18-9 | C8H14O2 | 142.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2497-18-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00009474
InChI Key HRHOWZHRCRZVCU-AATRIKPKSA-N
Synonym z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z
PubChem CID 2733294
IUPAC Name [(E)-hex-2-enyl] acetate
SMILES CCCC=CCOC(=O)C

Specifications

Density 0.9
Boiling Point 165°C to 166°C
Flash Point 58°C (136°F)
Refractive Index 1.427
Quantity 250 g
UN Number UN3272
Beilstein 1721851
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material trans-2-Hexenyl acetate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 219-680-7
RTECSNumber MP8425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.