Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Methyl-2-butenal, 97%

Catalog No. aal0811306
Click to view available options
Quantity:
5 g
25 g

CAS: 497-03-0 | C5H8O | 84.118 g/mol

Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-03-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00006977
InChI Key ACWQBUSCFPJUPN-HWKANZROSA-N
Synonym tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e
PubChem CID 5321950
IUPAC Name (E)-2-methylbut-2-enal
SMILES CC=C(C)C=O

Specifications

Density 0.871
Boiling Point 117°C to 119°C
Flash Point 18°C (64°F)
Odor Fruit-like
Refractive Index 1.448
Quantity 5 g
UN Number UN1989
Beilstein 1698207
Sensitivity Air sensitive
Solubility Information Soluble in water
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material trans-2-Methyl-2-butenal
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 207-833-0
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.