Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Methyl-2-butenal, 98%, Thermo Scientific Chemicals

Catalog Number AC353030050
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
497-03-0
C5H8O
84.12
MFCD00006977
ACWQBUSCFPJUPN-HWKANZROSA-N
tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e
5321950
(E)-2-methylbut-2-enal
CC=C(C)C=O
497-03-0
C5H8O
84.12
MFCD00006977
ACWQBUSCFPJUPN-HWKANZROSA-N
tiglic aldehyde, trans-2-methyl-2-butenal, tiglaldehyde, 2-methylbut-2-enal, e-2-methylbut-2-enal, tiglic acid aldehyde, e-2-methyl-2-butenal, 2-butenal, 2-methyl-, e, tigaldehyde, trans, 2-butenal, 2-methyl-, 2e
5321950
(E)-2-methylbut-2-enal
CC=C(C)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8710g/mL
Boiling Point 115°C to 118°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.447
Linear Formula CH3CH=C(CH3)CHO
Quantity 5 g
Beilstein 01, IV, 3464
Specific Gravity 0.871
Solubility Information Solubility in water: slightly soluble
Formula Weight 84.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material trans-2-Methyl-2-butenal, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 207-833-
Recommended Storage Flammables

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit