Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Nonenal, 95%, Thermo Scientific™

Catalog No. ac298310250
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle

Chemical Identifiers

CAS 18829-56-6
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00007012
InChI Key BSAIUMLZVGUGKX-BQYQJAHWSA-N
Synonym trans-2-nonenal, 2-nonenal, e-2-nonenal, e-non-2-enal, 3-hexylacrolein, 2-nonenal, 2e, beta-hexylacrolein, 2-nonenal, e, heptylideneacetaldehyde, alpha-nonenyl aldehyde
PubChem CID 5283335
IUPAC Name (E)-non-2-enal
SMILES CCCCCCC=CC=O

Specifications

Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 55°C to 60°C (0.3 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4520 to 1.456
Linear Formula CH3(CH2)5CH=CHCHO
Quantity 25 mL
Specific Gravity 0.84
Merck Index 15, 6763
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,fixed oils
Formula Weight 140.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material trans-2-Nonenal, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
EINECSNumber 242-609-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.