Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Octen-1-ol, 97%

Catalog No. p-7022955
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 18409-17-1 | C8H16O | 128.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18409-17-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00014057
InChI Key AYQPVPFZWIQERS-VOTSOKGWSA-N
Synonym trans-2-octen-1-ol, e-oct-2-en-1-ol, e-2-octen-1-ol, 2-octenol, 2-octen-1-ol, e, 2-octen-1-ol, 2e-octen-1-ol, unii-k54st7v77u, 2-octen-1-ol, 2e, 2e-oct-2-en-1-ol
PubChem CID 5318599
IUPAC Name (E)-oct-2-en-1-ol
SMILES CCCCCC=CCO

Specifications

Density 0.85
Boiling Point 88°C (11 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.4465
Quantity 5 g
Beilstein 1720707
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material trans-2-Octen-1-ol
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 242-288-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.