Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Pentenal, 97%

Catalog No. p-180056
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1576-87-0 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-87-0
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00009615
InChI Key DTCCTIQRPGSLPT-ONEGZZNKSA-N
Synonym trans-2-pentenal, 2-pentenal, e-2-pentenal, e-pent-2-enal, 3-ethylacrolein, pent-2-enal, 2-pentenal, e, 2-e-pentenal, 3-ethyl-2-propenal, e-pent-2-en-1-al
PubChem CID 5364752
IUPAC Name (E)-pent-2-enal
SMILES CCC=CC=O

Specifications

Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 115.0°C to 125.0°C
Flash Point 19°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4400 to 1.4470
Linear Formula CHOHC=CHC2H5
Quantity 5 g
Beilstein 01, IV, 3459
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble
Formula Weight 84.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material trans-2-Pentenal, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
EINECSNumber 216-414-1
RTECSNumber SB1560000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.