Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Pentenoic Acid 95.0+%, TCI America™
SDP

Catalog No. P034525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P034525ML 25 mL
P03455ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P034525ML Supplier TCI America Supplier No. P034525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13991-37-2
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00002704
InChI Key YIYBQIKDCADOSF-ONEGZZNKSA-N
Synonym trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
PubChem CID 638122
ChEBI CHEBI:38366
IUPAC Name (2E)-pent-2-enoic acid
SMILES CC\C=C\C(O)=O

Specifications

Color Yellow
Boiling Point 97°C
Quantity 25 mL
UN Number 3265
Formula Weight 100.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Pentenoic Acid
EINECSNumber (2)-0609
RTECSNumber SB2750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.