Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Phenyl-1-cyclopropanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. P075025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P075025G 25 g
P07505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P075025G Supplier TCI America Supplier No. P075025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 939-90-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00001292
InChI Key AHDDRJBFJBDEPW-DTWKUNHWSA-N
Synonym trans-2-phenylcyclopropanecarboxylic acid, trans-2-phenyl-1-cyclopropanecarboxylic acid, 1r,2r-2-phenylcyclopropanecarboxylic acid, trans-2-phenylcyclopropane-1-carboxylic acid, 2-phcpcooh, 1r,2r-2-phenylcyclopropane-1-carboxylic acid, cyclopropanecarboxylic acid, 2-phenyl-, 1r,2r-rel, rac trans-2-phenylcyclopropanecarboxylic-d5 acid, pubchem22341, trans-2-phenylcyclopropancarboxylic acid
PubChem CID 237413
ChEBI CHEBI:79920
IUPAC Name (1R,2R)-2-phenylcyclopropane-1-carboxylic acid
SMILES C1C(C1C(=O)O)C2=CC=CC=C2

Specifications

Melting Point 88°C
Quantity 25 g
Formula Weight 162.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2-Phenyl-1-cyclopropanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.