Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
trans-2-Tetradecenoyl-L-carnitine, Analytical Standard, ≥95.0% (LC/MS), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
10 mg
Specifications
Specifications
Linear Formula | C21H39NO4 |
Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[[(2 E)-1-oxo-2-tetradecen-1-yl]oxy]-1-propanaminium inner salt; trans-2-Myristenoyl-L-carnitine; C14:1-Carnitine |
Quantity | 10 mg |
Formula Weight | 369.54 |
Percent Purity | ≥95.0% (LC/MS) |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction