Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
trans-3-Bromocinnamic Acid 99.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 180°C |
Color | White |
Quantity | 25 g |
Formula Weight | 227.06 |
Percent Purity | ≥99.0% (GC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | trans-3-Bromocinnamic Acid |
Safety and Handling
RTECSNumber | GD8420000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction