Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-Hexen-1-ol, 97%

Catalog No. aal1054106
Click to view available options
Quantity:
5 g
25 g

CAS: 928-97-2 | C6H12O | 100.161 g/mol

trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-97-2
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00002960
InChI Key UFLHIIWVXFIJGU-ONEGZZNKSA-N
Synonym trans-3-hexen-1-ol, e-hex-3-en-1-ol, e-3-hexen-1-ol, trans-3-hexenol, 3-hexen-1-ol, 3e-hex-3-en-1-ol, 3-hexen-1-ol, e, trans-hex-3-en-1-ol, unii-4e0nfr5b3u, 3e-hexen-1-ol
PubChem CID 5284503
IUPAC Name (E)-hex-3-en-1-ol
SMILES CCC=CCCO

Specifications

Density 0.845
Boiling Point 60°C to 61°C (12 mmHg)
Flash Point 58°C (136°F)
Odor Pleasant
Refractive Index 1.439
Quantity 5 g
UN Number UN1987
Beilstein 1719713
Merck Index 14,4700
Solubility Information Slightly soluble in water.
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material trans-3-Hexen-1-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 213-193-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.