Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%

Catalog No. AAB2174230
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 1197-18-8 | C8H15NO2 | 157.213 g/mol

It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an antifibrinolytic agent that is found to inhibit plasmin-induced fibrinolysis. Used in various hemorrhagic diseases , abnormal bleeding in operations. It is also used as a lysine analogue to characterize binding sites in plasminogen.

Solubility
Soluble in water (167 mg/ml), ethanol (<1mg/mL at 25°C). Very slightly soluble in ether, DMSO (<1mg/mL at 25°C), and alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197-18-8
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00001466
InChI Key GYDJEQRTZSCIOI-UHFFFAOYSA-N
Synonym tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron
PubChem CID 5526
IUPAC Name 4-(aminomethyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1CN)C(=O)O

Specifications

Melting Point >300°C
Beilstein 2207452
Merck Index 14,9569
Solubility Information Soluble in water (167mg/ml),ethanol (<1mg/ml at 25°C). Very slightly soluble in ether,DMSO (<1mg/ml at 25°C),and alcohol.
Quantity 250 g
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-818-2
RTECSNumber GU8400000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.