Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Methoxy-beta-nitrostyrene, 98%

Catalog No. AAA1274514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1274514 25 g
AAA1274506 5 g
AAA1274522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1274514 Supplier Thermo Scientific Chemicals Supplier No. A1274514
Add to Cart

CAS: 5576-97-6 | C9H9NO3 | 179.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5576-97-6
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00024826
InChI Key JKQUXSHVQGBODD-UHFFFAOYSA-N
Synonym 1-methoxy-4-2-nitrovinyl benzene, 4-methoxy-beta-nitrostyrene, p-2-nitrovinyl anisole, trans-4-methoxy-beta-nitrostyrene, e-1-methoxy-4-2-nitrovinyl benzene, 2'-nitro vinyl anisole, 1-methoxy-4-e-2-nitroethenyl benzene, 1-4-methoxyphenyl-2-nitroethene, 1-4-methoxyphenyl-2-nitroethylene, 4-2-nitrovinyl anisole
PubChem CID 697963
SMILES COC1=CC=C(C=C[N+]([O-])=O)C=C1

Specifications

Melting Point 86°C to 88°C
Quantity 25 g
Beilstein 2047935
IUPAC Name 1-methoxy-4-[(E)-2-nitroethenyl]benzene
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material trans-4-Methoxy-beta-nitrostyrene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.