Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Methylcyclohexylamine, 99%

Catalog No. AC455920250
Click to view available options
Quantity:
25 g

CAS: 2523-55-9 | C7H15N | 113.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2523-55-9
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.2
InChI Key KSMVBYPXNKCPAJ-UHFFFAOYSA-N
Synonym 4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine
PubChem CID 80604
IUPAC Name 4-methylcyclohexan-1-amine
SMILES CC1CCC(CC1)N

Specifications

Density 0.8500g/mL
Boiling Point 151.0°C to 154.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Refractive Index 1.4500 to 1.5480
Linear Formula CH3C6H10NH2
Quantity 25 g
Specific Gravity 0.85
Formula Weight 113.2
Percent Purity 99%
Chemical Name or Material trans-4-Methylcyclohexylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Flammable liquid and vapour. Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.