Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Phenyl-3-buten-2-one, 99%

Catalog No. ac164520500
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Glass bottle

Catalog No. AC164520500

Supplier: Thermo Scientific Chemicals 164520500

Only null left
Add to Cart

CAS: 1896-62-4 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1896-62-4
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00008779
InChI Key BWHOZHOGCMHOBV-BQYQJAHWSA-N
Synonym benzalacetone, benzylideneacetone, methyl styryl ketone, 4-phenylbut-3-en-2-one, acetocinnamone, 4-phenyl-3-buten-2-one, e-4-phenylbut-3-en-2-one, benzylidene acetone, trans-benzalacetone, benzalaceton
PubChem CID 637759
ChEBI CHEBI:78399
IUPAC Name (E)-4-phenylbut-3-en-2-one
SMILES CC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 38.5°C to 41°C
Color Yellow
Boiling Point 260°C to 262°C
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCOCH3
Quantity 50 g
Beilstein 07, 364
Merck Index 15, 1140
Solubility Information Solubility in water: negligible. Other solubilities: freely soluble in alcohol,benzene,chloroform and,ether
Formula Weight 146.19
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material trans-4-Phenyl-3-buten-2-one, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice
EINECSNumber 217-587-6
RTECSNumber EN0330050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.