Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-5-Decene, 97%

Catalog No. p-7045429
Click to view available options
Quantity:
1 g
5 g

CAS: 7433-56-9 | C10H20 | 140.27 g/mol

trans-5-Decene has been used to characterize the effect of cyclization and size on the stability of Criegee intermediates formed in ozonolysis. It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-5-Decene has been used to characterize the effect of cyclization and size on the stability of Criegee intermediates formed in ozonolysis. It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7433-56-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00009508
InChI Key UURSXESKOOOTOV-MDZDMXLPSA-N
Synonym trans-5-decene, e-5-decene, 5-decene, e, trans-dec-5-ene, 5e-decene, 5-decene, cis-5-decene, 5-decene, 5e, 5-trans-decene, 5e dec-5-ene
PubChem CID 637820
IUPAC Name (E)-dec-5-ene
SMILES CCCC\C=C\CCCC

Specifications

Density 0.74
Boiling Point 171°C to 172°C
Flash Point 46°C (114°F)
Refractive Index 1.424
Quantity 1 g
UN Number UN3295
Beilstein 1719487
Solubility Information Insoluble in water.
Formula Weight 140.27
Percent Purity 97%
Chemical Name or Material trans-5-Decene
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 231-080-7
RTECSNumber HE2080000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.