Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Anethole, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187576 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL

trans-Anethole is a naturally occuring flavouring agent. It has insecticidal, larvicidal, and antimicrobial properties.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 4180-23-8
Solution Contains ˜0.05% 4-tert-butylcatechol as stabilizer
Density 0.988 g/mL (at 25°C)
Flash Point 91°C
Physical Form Neat
Linear Formula CH3 CH=CHC6H4 OCH3
MDL Number MFCD00009284
Melting Point 20°C to 21°C
Boiling Point 234°C to 237°C
Synonym 4-Propenylanisole; trans-1-Methoxy-4-(1-propenyl)benzene
Type Analytical Standard
Quantity 1 mL
Formula Weight 148.20
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.