Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-trans-Azetidine-2,4-dicarboxylic acid, 96%, Thermo Scientific™

Catalog No. AC329010010
Click to view available options
Quantity:
1 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121050-03-1
Molecular Formula C5H7NO4
Molecular Weight (g/mol) 145.114
MDL Number MFCD00236469
InChI Key JMVIGOFRIJJUAW-PWNYCUMCSA-N
Synonym 2r,4r-azetidin-1-ium-2,4-dicarboxylate
PubChem CID 124432
IUPAC Name (2R,4R)-azetidine-2,4-dicarboxylic acid
SMILES C1C(NC1C(=O)O)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Quantity 1 mg
Formula Weight 145.1
Physical Form Powder
Chemical Name or Material ±-trans-Azetidine-2, 4-dicarboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.