Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Dichlorobis(diethylsulfide)platinum(II), Pt 43.7%

Catalog No. p-7048417
Click to view available options
Quantity:
1 g
5 g

Hydrosilation | CAS: 15337-84-5 | C8H20Cl2PtS2 | 446.35 g/mol

It is used in Hydrosilylation. Platinum(II) complexes of diethylsulfide are convenient and versatile precursor for the preparation of a variety of organoplatinum(II) derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15337-84-5
Molecular Formula C8H20Cl2PtS2
Molecular Weight (g/mol) 446.35
MDL Number MFCD00050943
InChI Key RFSLRYGKHBDFGC-UHFFFAOYSA-L
IUPAC Name platinum(2+) bis((ethylsulfanyl)ethane) dichloride
SMILES Cl[Pt++]Cl.CCSCC.CCSCC

Specifications

Quantity 1 g
Solubility Information Soluble in acetone,alcohol,ethyl acetate and benzene
Formula Weight 446.37
Concentration or Composition (by Analyte or Components) Pt 43.7%
Physical Form Crystalline
Chemical Name or Material trans-Dichlorobis(diethylsulfide)platinum(II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-373-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.