Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ trans-ISRIB
GREENER_CHOICESDP

Catalog No. 528410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-841-0 10 mg
52-845-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-841-0 Supplier Tocris Bioscience™ Supplier No. 5284/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Integrated stress response (ISR) inhibitor

trans-ISRIB is a potent integrated stress response (ISR) inhibitor; reverses the effects of eIF2α phosphorylation (IC50 = 5 nM) and restores cell translation capacity. Acts downstream of PERK, targeting interactions between eIF2α kinases and elF2B. Blocks ATF4 production without altering the IRE1 or ATF6 responses in vitro. Displays memory enhancement in hippocampus-dependent spatial and fear-associated learning in rodents. trans-ISRIB promotes survival of pluripotent stem cells in culture when used in combination with Chroman 1 (Cat. No. 7163), Emricasan (Cat. No. 7310), and Polyamine Supplement x1000 (lyophilized) (Cat. No. 7739), a combination known as CEPT. When used as part of the CEPT cocktail, trans-ISRIB improves survival of differentiated cells following cryopreservation. Brain penetrant.

Specifications

Quantity 10 mg
Product Type trans-ISRIB
Purity >99%
Molecular Weight (g/mol) 451.34
Inhibitors trans-ISRIB

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.