Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-N-Benzyloxycarbonyl-4-tert-butoxy-L-proline, 95%, Thermo Scientific™

Catalog No. AAH6503906
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85201-91-8
Molecular Formula C17H23NO5
Molecular Weight (g/mol) 321.373
MDL Number MFCD00153336
InChI Key YCTNIMOEFJORHQ-KGLIPLIRSA-N
Synonym z-hyp tbu-oh, z-o-tert-butyl-l-4-hydroxyproline, 2s,4r-4-tert-butoxy-1,2-pyrrolidinedicarboxylic acid 1-benzyl ester, 2s,4r-4-tert-butoxy-pyrrolidine-1,2-dicarboxylic acid 1-benzyl ester, 2s,4r-1-benzyloxy carbonyl-4-tert-butoxy pyrrolidine-2-carboxylic acid, 2s,4r-4-2-methylpropan-2-yl oxy-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
PubChem CID 6950951
IUPAC Name (2S,4R)-4-[(2-methylpropan-2-yl)oxy]-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Beilstein 5088264
Quantity 5 g
Formula Weight 321.37
Percent Purity 95%
Physical Form Solid
Chemical Name or Material trans-N-Benzyloxycarbonyl-4-tert-butoxy-L-proline
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.