Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-Stilbene oxide, 99%, Thermo Scientific Chemicals

Catalog No. AC132780050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132780050 5 g Glass bottle
AC132780010 1 g Glass bottle
AC132780250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC132780050 Supplier Thermo Scientific Chemicals Supplier No. 132780050
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1439-07-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00064311
InChI Key ARCJQKUWGAZPFX-KWCCSABGSA-N
Synonym trans-stilbene oxide, 3r-2,3-diphenyloxirane, 2r-2alpha,3-diphenyloxirane
PubChem CID 5742860
IUPAC Name (3R)-2,3-diphenyloxirane
SMILES C1=CC=C(C=C1)C2C(O2)C3=CC=CC=C3

Specifications

Melting Point 67°C to 70°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 17, I, 31
Formula Weight 196.25
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material trans-Stilbene oxide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-877-7
RTECSNumber DT4391500

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.